missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543960
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: DVWKELSLLL QVEQRMPVSP ISQGASWAQE DQQDADEDRR AFQMLRRDNE Sequence homology: Cow: 85%; Horse: 91%; Human: 100%; Rat: 90%
MLKL Polyclonal antibody specifically detects MLKL in Human samples. It is validated for Western Blot.Specifications
MLKL | |
Polyclonal | |
Unconjugated | |
Mlkl | |
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
197259 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NB16 | |
Mlkl | |
Synthetic peptide directed towards the N-terminal of human MLKL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |