missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533917
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Mouse, Hamster, Rabbit, Pig (100%).
MLK1 Polyclonal antibody specifically detects MLK1 in Human, Mouse, Porcine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Specifications
MLK1 | |
Polyclonal | |
Unconjugated | |
MAP3K9 | |
E130314H24Rik; Map3k9; MEKK9; mitogen activated protein kinase kinase kinase 9; mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); mixed-lineage kinase 1; MLK1; Prke1; RGD1562149 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100155753, 100341731, 338372, 4293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P80192, Q3U1V8 | |
MAP3K9 | |
Synthetic 15 amino acid peptide from internal region of human MAP3K9. | |
50 μg | |
Primary | |
Human, Mouse, Porcine, Rabbit | |
Antibody | |
IgG |