Learn More
MKNK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544322
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SDFGLQCSAR PDMPASQPID IPDAKKRGKK KKRGRATDSF SGRFEDVYQL Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 77%; Rat: 100%; Zebrafish: 100%
MKNK2 Polyclonal antibody specifically detects MKNK2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MKNK2 | |
Polyclonal | |
Unconjugated | |
Mknk2 | |
2010016G11Rik; G protein-coupled receptor kinase 7; GPRK7; MAP kinase interacting serine/threonine kinase 2; MAP kinase signal-integrating kinase 2; MAP kinase-interacting serine/threonine kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; LOW QUALITY PROTEIN: MAP kinase-interacting serine/threonine-protein kinase 2; MAPK interacting serine/threonine kinase 2; MAPK signal-integrating kinase 2; Mknk2; MNK2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2872 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HBH9 | |
Mknk2 | |
Synthetic peptide directed towards the N-terminal of human MKNK2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |