Learn More
MiTF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529826
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, mouse skin. Predicted reactivity: Mouse (92%), Rat (92%), Dog (95%), Pig (99%), Chicken (93%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MiTF Polyclonal antibody specifically detects MiTF in Human, Mouse samples. It is validated for Western Blot.Specifications
MiTF | |
Polyclonal | |
Unconjugated | |
MITF | |
BCC2; Bhlhe32; black eyed white; bw; Class E basic helix-loop-helix protein 32; CMM8; Gsfbcc2; melanocyte inducing transcription factor; melanogenesis associated transcription factor; mi; microphtalmia-associated transcription factor; microphthalmia; microphthalmia associated transcription factor isoform M-; microphthalmia transcription factor; microphthalmia-associated transcription factor; microphthalmia-associated transcription factor A; microphthalmia-associated transcription factor isoform H; microphthalmia-associated transcription factor isoform H protein; microphthalmia-associated transcription factor Trnc-1; microphthalmia-associated transcription factor Trnc-2; microphthalmia-associated transcription factor variant 1; microphthalmia-associated transcription factor variant 2; microphthalmia-associated transcription factor variant 3; microphthalmia-associated transcription factor variant MITF-M(+); MITF; MITF-H; MITF-M; MITF-M-; transcription factor, isoform 7; vit; Vitiligo; Wh | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17342, 4286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O75030, Q08874 | |
MITF | |
Recombinant protein fragment corresponding to a region within amino acids 42 and 191 of Human MITF | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |