Kimberly Clark™ Anti mist Platinum eyewear.
Overview
Brand: Kimberly Clark™
Manufacturer Part Number: 25710
X12 EYEWEAR PLATINIUM X V40 SMOKY
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 12611416
 N/A
N/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description and Specification
Specification
Quantity 12/Pack
Product Type Safety Eyewear
Lens Type Anti mist
For Use With (Application) Reducing glare, eye strain and fatigue while working in bright sunlight, keeping lenses clear and free from mist in damp areas or during temperature or humidity changes
Certifications/Compliance EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.