Learn More
MIP-1 gamma Monoclonal Antibody (62105), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA523840
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In sandwich ELISAs, less than 0.02% cross-reactivity with recombinant human CCL3, 4, 15, 19, 20, recombinant mouse CCL3, 4, 19, or recombinant rat CCL20 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
MIP-1 gamma Monoclonal antibody specifically detects MIP-1 gamma in Mouse samples. It is validated for ELISA, Neutralization, Western Blot.Specifications
MIP-1 gamma | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P51670 | |
Ccl9 | |
E. coli-derived recombinant mouse CCL9/10/MIP-1 gamma Gln22-Gln122 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
ELISA, Neutralization, Western Blot | |
62105 | |
Unconjugated | |
Ccl9 | |
C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9 | |
Rat | |
Protein A/G | |
RUO | |
20308 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |