Learn More
MINA53 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531229
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Human (99%), Rat (80%), Rhesus Monkey (98%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MINA53 Polyclonal antibody specifically detects MINA53 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MINA53 | |
Polyclonal | |
Unconjugated | |
RIOX2 | |
1810047J07Rik; 2410057H13Rik; 3830408E23Rik; 60S ribosomal protein L27a histidine hydroxylase; AI449204; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; DKFZp762O1912; FLJ14393; histone lysine demethylase MINA; MDIG; mina; mina {ECO:0000312; Mina53; Mina-53; mineral dust induced gene protein; mineral dust-induced gene protein; MYC induced nuclear antigen; myc-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; NO52; Nucleolar protein 52; Protein mina53; RGD:708521}; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; RIOX2; ROX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IUF8 | |
RIOX2 | |
Recombinant protein fragment corresponding to a region within amino acids 105 and 409 of Human MINA53 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |