Learn More
Mimecan Rat anti-Mouse, Clone: 329939, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2949
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429805
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Mimecan Monoclonal antibody MAB2949 Specifically detects Mimecan in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
Mimecan/OIF
329939
Western Blot 1 ug/mL
Q62000
DKFZp586P2421, KSPG25, mimecan, mimecan proteoglycan, OGN, OIF, OIFOG, osteoglycin, osteoglycin (osteoinductive factor), Osteoinductive factor, SLRR3A, SLRR3Acorneal keratan sulfate proteoglycan
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Mimecan in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Mimecan is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
OGN
Mouse myeloma cell line NS0-derived recombinant mouse Mimecan Ala20-Phe298 Accession # Q62000
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4969
Reconstitute at 0.5 mg/mL in sterile PBS.