Learn More
Kindlin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531168
Code : PN
Additional Details : Weight : 0.01000kg
Description
PA5-31168 targets Mig-2 in IHC (P) and WB applications and shows reactivity with Human and Rat samples. The PA5-31168 immunogen is recombit protein fragment corresponding to a region within amino acids 92 and 680 of Human Mig-2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Kindlin 2 | |
Polyclonal | |
Unconjugated | |
Q96AC1 | |
FERMT2 | |
Recombit protein fragment corresponding to a region within amino acids 92 and 680 of Human Mig-2 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
FERMT2 | |
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2, PH domain-containing family C member 1, fermitin family homolog 2, kindlin 2, kindlin-2, mitogen inducible gene 2 protein, mitogen-inducible gene 2 protein, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain c | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10979, 289992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |