Learn More
HLA-DR/DP/DQ Monoclonal Antibody (WR18), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180680
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 microliter of the suggested working dilution to label 1x10^6 cells in 100 microliter. Mouse anti Human HLA DP DQ DR antibody, clone WR18 reacts with a monomorphic determinant common to DP, DQ and DR beta chains, which are expressed by antigen presenting cells, B cells, monocytes and activated T lymphocytes.
HLA-DR/DP/DQ Monoclonal antibody specifically detects HLA-DR/DP/DQ in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DR/DP/DQ | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P01911, P01920, P04440 | |
HLA-DPB1, HLA-DQB1, HLA-DRB1 | |
Human HLA Class II (DP,DQ,DR). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
WR18 | |
PE | |
HLA-DQB1 | |
beta1 domain MHC class II HLA DPB; CELIAC1; class II HLA beta chain; Clone P2-beta-3; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DPB1; DR-16; DRB; DRB1; DRBB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II antigen; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQ; HLA-DQB; HLA-DQB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; IDDM1; integral membrane glycoprotein; LA-DRB; lymphocyte antigen; lymphocyte a | |
Mouse | |
Protein G | |
RUO | |
3115, 3119, 3123 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |
Safety and Handling
- HLA-DR/DP/DQ Monoclonal Antibody (WR18), PE
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H302-Harmful if swallowed.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide