missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC II Monomorphic Monoclonal Antibody (CA2.1C12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516608
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
MHC II Monomorphic Monoclonal antibody specifically detects MHC II Monomorphic in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
MHC II Monomorphic | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P18470 | |
DLA-DQA1, DLA-DQB1, HLA-DRB1 | |
The details of the immunogen for this antibody are not currently available. | |
RUO | |
474860, 474861, 474862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
CA2.1C12 | |
Unconjugated | |
DLA-DQB1 | |
beta 1 domain; B-L; B-L beta chain; B-L beta minor; BLA-DRB3; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; BoLA-DQA; BoLA-DQA1; BOLA-DQA2; BOLA-DQA2 protein; BOLA-DQA3; BOLA-DQA5; BOLA-DQB; BOLA-DRA; BoLA-DR-alpha; BoLA-DRB; BOLA-DRB3; BoLA-DRB3 protein; BoLA-DRB3.2; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; bovine leukocyte antigen; class II histocompatibility antigen, B-L beta chain; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DQA; DLA-DQA1; DLA-DQB; DLA-DQB1; DLA-DR beta; DLA-DRB1; DLA-DRBB1; dog leucocyte antigen DQA; DQA; dqa 7; dqa1; DQALPHA; DQB; dqb1; DR beta-chain antigen binding domain; DRA; DRB; DRB1; DRB3; DRBB1; DR-beta chain; DR-beta chain MHC class II; ELA-DQA; ela-DRA; Eqca-DQA1; Eqca-DQA2; Eqca-DQA3; Eqca-DQB1; Eqca-DQB2; Eqca-DRA; Eqca-DRB1; Eqca-DRB2; Eqca-DRB3; GSP-BLB1; GSP-BLB2; histocompatibility complex, class II, DR beta 3; HLA-DRB1; integral mem | |
Mouse | |
2 mL | |
Primary | |
Canine | |
Antibody | |
IgG1 |