Learn More
mGluR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533821
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Mouse, Rabbit, Pig (100%) Rat, Hamster, Dog (94%) Horse, Opossum (88%).
mGluR4 Polyclonal antibody specifically detects mGluR4 in Human, Mouse, Non-human Primate, Rabbit samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
mGluR4 | |
Polyclonal | |
Unconjugated | |
GRM4 | |
G protein-coupled receptor GRM4; G protein-coupled receptor, family C, group 1, member D; glutamate metabotropic receptor 4; glutamate receptor; glutamate receptor, metabotropic 4; GPRC1D; GRM4; Metabotropic glutamate receptor 4; mGlu4; MGLUR4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100347907, 268934, 2914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14833, Q68EF4 | |
GRM4 | |
Synthetic 16 amino acid peptide from C-terminus of human GRM4 / MGLUR4. | |
50 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rabbit | |
Antibody | |
IgG |