missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545103
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ARDLADVAAR EAEREGATEE AVRERRETEL CEHWEEHTRQ RELPLRGPLG Sequence homology: Human: 100%
MRI1 Polyclonal antibody specifically detects MRI1 in Human samples. It is validated for Western Blot.Specifications
MRI1 | |
Polyclonal | |
Unconjugated | |
MRI1 | |
2410018C20Rik; CH73-131E21.1; HAMAP-Rule:MF_03119}; M1Pi; M1Pi {ECO:0000255; mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase {ECO:0000255; methylthioribose-1-phosphate isomerase 1; methylthioribose-1-phosphate isomerase homolog; methylthioribose-1-phosphate isomerase homolog (S. cerevisiae); MGC3207; MRDI; Mri1; MTNA; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255; RGD1307789; similar to CG11334-PB, isoform B; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255; S-methyl-5-thioribose-1-phosphate isomerase 1; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255; UNQ6390/PRO21135; Ypr118w; zgc:172216 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84245 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BV20 | |
MRI1 | |
Synthetic peptide directed towards the N-terminal of human MGC3207 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |