Learn More
MFN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527606
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MFN2-transfected 293T. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (83%), Xenopus laevis (82%), Dog (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MFN2 Polyclonal antibody specifically detects MFN2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MFN2 | |
Polyclonal | |
Unconjugated | |
MFN2 | |
CMT2A; CMT2A2; CPRP 1; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; hypertension-related protein 1; Kiaa0214; LOW QUALITY PROTEIN: mitofusin-2; Marf; MFN 2; MFN2; mg:cb01g09; Mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; mitofusin 2; Mitofusin2; mitofusin-2; si:dkeyp-104h9.2; Transmembrane GTPase MFN2; wu:fb79a11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170731, 64476, 9927 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95140, Q80U63, Q8R500 | |
MFN2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 183 of Human MFN2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |