missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSRA Mouse anti-Human, Rat, Clone: 5B5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0084
Additional Details : Weight : 0.99000kg
Description
A suggested positive control for this product is HepG2 cells.
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer′s disease in mammals.Specifications
MSRA | |
Monoclonal | |
Unconjugated | |
Q9UJ68, Q923M1 | |
MSRA | |
Purified, recombit, human methionine sulfoxide reductase A protein expressed in E. coli. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
5B5 | |
MSRA | |
cytosolic methionine-S-sulfoxide reductase | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
29447, 4482 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |