Learn More
c-Met Recombinant Rabbit Monoclonal Antibody (22H22L13), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 700261
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with primate based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
c-Met Recombinant Monoclonal antibody specifically detects c-Met in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
c-Met | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AI838057; AUTS9; bHLHe59; CG1705; CG1705-PA; CG1705-PB; cmet; c-met; c-met proto-oncogene; c-Met receptor tyrosine kinase; c-Met/HGF receptor; D249; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; HGFR; HGF-SF receptor; juvenile hormone resistance; met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; Met; protein tyrosin kinase; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; Proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; scatter factor receptor; SF receptor; soluble c met; tyrosine-protein kinase Met | |
A peptide corresponding to amino acids 1379-1390 of P08581. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
22H22L13 | |
Unconjugated | |
P08581 | |
Rabbit | |
Protein A | |
RUO | |
4233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |