Learn More
Mesothelin Propeptide/MPF Mouse anti-Human, Clone: 346633, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3265
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469975
  • £320.46 / 100µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Mesothelin Propeptide/MPF Monoclonal antibody MAB3265 Specifically detects Mesothelin Propeptide/MPF in Human samples. It is validated for Western Blotting
Specifications
Specifications
Mesothelin
346633
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MSLN
Mouse myeloma cell line NS0-derived recombinant human Mesothelin propeptide Arg35-Leu289 Accession # AAH09272
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10232
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
AAH09272
CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Mesothelin Propeptide/MPF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Mesothelin is observed.