Learn More
Pro Meprin beta Subunit/MEP1B Rat anti-Human, Clone: 289703, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB28952
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429765
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Pro Meprin beta Subunit/MEP1B Monoclonal antibody MAB28952 Specifically detects Pro Meprin beta Subunit/MEP1B in Human samples. It is validated for Western Blotting
Specifications
Specifications
Meprin beta Subunit/MEP1B
289703
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MEP1B
Human embryonic kidney cell line HEK293-derived recombinant human Meprin beta Subunit/MEP1B Thr23-Ser593 Accession # Q16820
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4225
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q16820
EC 3.4.24.63, endopeptidase-2, MEP1B, meprin A subunit beta, meprin A, beta, Meprin B, N-benzoyl-L-tyrosyl-p-amino-benzoic acid hydrolase beta subunit, N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit beta, PABA peptide hydrolase, PPH beta
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human or mouse Pro Meprin beta Subunit/MEP1B in Western blots. It does not react with the mature form.