missing translation for 'onlineSavingsMsg'
Learn More

MEPE/OF45 Mouse anti-Human, Clone: 341611, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3140

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459915

  • £271.93 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

MEPE/OF45 Monoclonal antibody MAB3140 Specifically detects MEPE/OF45 in Human samples. It is validated for Western Blotting
Specifications

Specifications

MEPE/OF45
341611
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MEPE
Mouse myeloma cell line NS0-derived recombinant human MEPE/OF45, Pro18-Asp525, Accession # Q9NQ76
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
56955
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9NQ76
extracellular phosphoglycoprotein with ASARM motif (bone), matrix extracellular phosphoglycoprotein, OF45, Osteoregulin
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human MEPE/OF45 in direct ELISAs and Western blots.
Human
Videos

For Research Use Only