missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEPCE Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518813
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
MEPCE Polyclonal antibody specifically detects MEPCE in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MEPCE | |
Polyclonal | |
Unconjugated | |
mepce | |
7SK snRNA methylphosphate capping enzyme; BCDIN3; Bicoid-interacting protein 3 homolog; bin3 homolog; bin3, bicoid-interacting 3; bin3, bicoid-interacting 3, homolog; Bipl1; D5Wsu46e; fc59f06; FLJ20257; Mepce; methylphosphate capping enzyme; si:ch211-283h6.1; wu:fc59f06; zBCDIN3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
304361, 56257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q7L2J0 | |
mepce | |
Synthetic peptide sequence (KQQRKFQYGNYCK) corresponding to the internal amino acids of MEPCE | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |