missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525432
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
MEIS2 Polyclonal antibody specifically detects MEIS2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MEIS2 | |
Polyclonal | |
Unconjugated | |
MEIS2 | |
A430109D20Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; homeobox protein Meis2; HsT18361; Meis homeobox 2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; meis1-related protein 1; MEIS2; Meis2 homeobox; MGC2820; MRG1; myeloid ecotropic viral integration site-related gene 1; OTTHUMP00000160209; Stra10; TALE homeobox protein Meis2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O14770 | |
MEIS2 | |
KLH conjugated synthetic peptide between 254-280 amino acids from the central region of human MEIS2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |