missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD146 Monoclonal Antibody (404722), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524142
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) MCAM, rhBCAM, or rhALCAM is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD146 Monoclonal antibody specifically detects CD146 in Rat samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD146 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9EPF2 | |
MCAM | |
Mouse myeloma cell line NS0-derived recombinant rat MCAM/CD146 Arg19-Lys560 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
404722 | |
Unconjugated | |
MCAM | |
1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
Mouse | |
Protein A/G | |
RUO | |
78967 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |