missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MC5R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533929
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey, Panda (100%) Marmoset, Mouse, Dog, Bat, Hamster, Rabbit (94%).
MC5R Polyclonal antibody specifically detects MC5R in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
MC5R | |
Polyclonal | |
Unconjugated | |
MC5R | |
H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P33032 | |
MC5R | |
Synthetic 16 amino acid peptide from 1st extracellular domain of human MC5 Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |