Learn More
MBL Mouse anti-Human, Clone: 285623, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB23071
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449345
  • £390.72 / 500µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
MBL Monoclonal antibody MAB23071 Specifically detects MBL in Human samples. It is validated for Blocking (Receptor-ligand)
Specifications
Specifications
MBL
285623
Blockade of Receptor-ligand Interaction
AAH96182
COLEC1, COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBL2, MBL2D, MBLmannan-binding lectin, MBP, MBP1, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human MBL in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse MBL-2 is observed.
Human
Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MBL2
Mouse myeloma cell line NS0-derived recombinant human MBL Glu21-Ile248 Accession # AAH96182
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4153
Reconstitute at 0.5 mg/mL in sterile PBS.