Learn More
MBL-2 Rat anti-Mouse, Clone: 272801, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2208
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449265
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
MBL-2 Monoclonal antibody MAB2208 Specifically detects MBL-2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
MBL-2
272801
Western Blot 1 ug/mL
P41317
L-MBP, mannose-binding protein C, L-MBP, mannose-binding lectin (protein C) 2, MBL, MBL-C, MBP-C
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse MBL-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MBL is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mbl2
Mouse myeloma cell line NS0-derived recombinant mouse MBL-2 Glu19-Asp244 Accession # P41317
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
17195
Reconstitute at 0.5 mg/mL in sterile PBS.