Learn More
Matrilin-4 Mouse anti-Human, Mouse, Clone: 401127, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4205
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440456
  • £249.49 / 100µg
Estimated Shipment
08-07-2019
Description
Description
Matrilin-4 Monoclonal antibody MAB4205 Specifically detects Matrilin-4 in Human, Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Matrilin-4
401127
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
MATN4
Mouse myeloma cell line NS0-derived recombinant human Matrilin-4 Gln20-Asp499 (predicted) Accession # NP_085095
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8785
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_085095
FLJ14417, MATN4, matrilin 4, matrilin-4, Matrilin4
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse Matrilin-4 in direct ELISAs and Western blots.
Human, Mouse