Learn More
Marapsin/Pancreasin Rat anti-Mouse, Clone: 246406, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB19891
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459085
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Marapsin/Pancreasin Monoclonal antibody MAB19891 Specifically detects Marapsin/Pancreasin in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
Marapsin/Pancreasin
246406
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PRSS27
Mouse myeloma cell line NS0-derived recombinant mouse Marapsin/Pancreasin Ala23-Thr290 Accession # Q8BJR6
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
83886
Sterile PBS to a final concentration of 0.5 mg/mL.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q8BJR6
CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Marapsin in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human Marapsin or recombinant mouse Prostasin was observed.