Learn More
Marapsin/Pancreasin Mouse anti-Human, Clone: 243305, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1988
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439085
  • £376.32 / 500µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Marapsin/Pancreasin Monoclonal antibody MAB1988 Specifically detects Marapsin/Pancreasin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Marapsin/Pancreasin
243305
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PRSS27
Mouse myeloma cell line NS0-derived recombinant human Marapsin/Pancreasin Ala23-Lys290 Accession # Q9BQR3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
83886
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q9BQR3
CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Marapsin/Pancreasin in direct ELISAs and Western blots. Reacts with an epitope within the mature chain. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Marapsin is observed.
Human
Videos
Dokumentation
For Research Use Only