missing translation for 'onlineSavingsMsg'
Learn More

LC3B, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSM00026W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11584853

  • £257.00 / 150ĀµL


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Reconstitute with 150 ul of distilled water.
Specifications

Specifications

Map1lc3b
Polyclonal
whole serum with no preservative
Q9GZQ8
MAP1A/MAP1B LC3 B, Map1lc3b
Rabbit
150μL
Antibody
Polyclonal
Human
ImmunohistochemistryĆ¾Western Blot
Unconjugated
MAP1LC3B
Serum
MAP1LC3B
A synthetic peptide from human microtubule-associated proteins 1A/1B light chain 3B
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
81631
Videos