missing translation for 'onlineSavingsMsg'
Learn More

LC3B, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSM00025W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11574853

  • £257.00 / 150µL


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Reconstitute with 150 ul of distilled water.
Specifications

Specifications

Map1lc3b
Polyclonal
whole serum with no preservative
Q9CQV6, Q62625
MAP1A/MAP1B LC3 B, Map1lc3b
Rabbit
150μL
Antibody
Polyclonal
MurineþRat
ImmunohistochemistryþWestern Blot
Unconjugated
Map1lc3b
Serum
MAP1LC3B
A synthetic peptide from mouse microtubule-associated proteins 1A/1B light chain 3B
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
67443
Videos