Learn More
LC3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA118293
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
LC3B Polyclonal antibody specifically detects LC3B in Human, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
LC3B | |
Polyclonal | |
Unconjugated | |
MAP1LC3B2 | |
1010001C15Rik; Atg8; ATG8E; ATG8F; ATG8G; Autophagy-related protein LC3 B; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3 beta; LC3A; LC3B; light chain 3; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; MAP1LC3; MAP1LC3A; MAP1LC3B; MAP1LC3B2; MAP1LC3B-a; microtubule associated protein 1 light chain 3 beta; microtubule associated protein 1 light chain 3 beta 2; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3 beta 2; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3B-like; Microtubule-associated proteins 1A/1B light chain 3B-like protein; Mpl3; RP11-346K17.1; wu:fb60g11; zbs559; zgc:56434 | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
A6NCE7, Q62625, Q9GZQ8 | |
MAP1LC3B, MAP1LC3B2 | |
Synthetic peptide corresponding to the N-terminal residues of human MAP1LC3 B conguate to Blue Carrier Protein. | |
RUO | |
100359928, 643246, 64862, 81631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |