missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Polyclonal Antibody, APC, Invitrogen™
Goat Polyclonal Antibody
Brand: Thermo Scientific PA546879
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human MMR is observed.
CD206 Polyclonal antibody specifically detects CD206 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD206 | |
Polyclonal | |
Buffered saline solution with BSA and 0.09% sodium azide | |
Q61830 | |
Mrc1 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Ala1388 | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
APC | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
17533 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |