Learn More
M-CSF Mouse anti-Human, Clone: 26786, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2161
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459225
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
M-CSF Monoclonal antibody MAB2161 Specifically detects M-CSF in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
Specifications
M-CSF
26786
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells
Purified
CSF1
E. coli-derived recombinant human M-CSF
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1435
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human M-CSF.