Learn More
LZIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527630
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299. Predicted reactivity: Rhesus Monkey (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LZIP Polyclonal antibody specifically detects LZIP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
LZIP | |
Polyclonal | |
Unconjugated | |
CREB3 | |
AU044960; AW538053; basic leucine zipper protein; C80076; cAMP responsive element binding protein 3; cAMP-responsive element-binding protein 3; CREB3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; cyclic AMP-responsive element-binding protein 3; leucin zipper proitein; Leucine zipper protein; Luman; LZIP; LZIP-1; LZIP-2; N-terminal Luman; Processed cyclic AMP-responsive element-binding protein 3; sLZIP; small leucine zipper protein; transcription factor LZIP; transcription factor LZIP-alpha | |
Rabbit | |
Protein A | |
RUO | |
10488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
O43889 | |
CREB3 | |
Recombinant fragment corresponding to a region within amino acids 231 and 395 of Human LZIP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |