Learn More
LYVE-1 Mouse anti-Human, Clone: 264725, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB20891
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459165
  • £364.33 / 500µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
LYVE-1 Monoclonal antibody MAB20891 Specifically detects LYVE-1 in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
LYVE-1
264725
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LYVE1
Mouse myeloma cell line NS0-derived recombinant human LYVE-1 Ser24-Thr238 Accession # Q9Y5Y7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10894
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9Y5Y7
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human LYVE-1 in ELISAs and Western blots.