missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAP2C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112679
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is transfected HEK-293 cells.
PPAP2C Polyclonal antibody specifically detects PPAP2C in Human samples. It is validated for Western Blot.Specifications
PPAP2C | |
Polyclonal | |
PBS with 0.08% sodium azide | |
O43688 | |
PLPP2 | |
Synthetic peptide corresponding to human lysophospholipid phosphatase 2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PLPP2 | |
lipid phosphate phosphohydrolase 2; LPP2; LPR3; PAP2c; PAP-2c; PAP2-G; PAP2-gamma; phosphatidate phosphohydrolase type 2c; Phosphatidic acid phosphatase 2c; phosphatidic acid phosphatase type 2C; phosphatidic acid phosphohydrolase type 2c; phospholipid phosphatase 2; PLPP2; Ppap2c; PRG2; PRG-2; type-2 phosphatidic acid phosphatase-gamma | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
8612 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |