missing translation for 'onlineSavingsMsg'
Learn More

Lysine Hydroxylase 2/PLOD2 Mouse anti-Human, Clone: 408105, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4445

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460566

  • £271.93 / 100µg


Estimated Shipment
30-10-2018
Add to basket

Description

Description

Lysine Hydroxylase 2/PLOD2 Monoclonal antibody MAB4445 Specifically detects Lysine Hydroxylase 2/PLOD2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Lysine Hydroxylase 2/PLOD2
408105
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PLOD2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lysine Hydroxylase 2/PLOD2, Ala26-Pro758, Accession # NP_891988
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5352
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_891988
2-oxoglutarate 5-dioxygenase (lysine hydroxylase) 2, 2-oxoglutarate 5-dioxygenase 2, LH2, PLOD2, procollagen-lysine, telopeptide lysyl hydroxylase, TLH
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Lysine Hydroxylase 2/PLOD2 in direct ELISAs and Western blots.
Human
Videos

For Research Use Only