missing translation for 'onlineSavingsMsg'
Learn More

anti-LXR alpha/NR1H3, Clone: K8607, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) PP-K8607-10

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13442456

  • £408.22 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications:

Direct ELISA, Gel Supershift, Immunoprecipitation, Western Blotting

Specifications

Specifications

LXR alpha/NR1H3
K8607
Western Blot, Immunoprecipitation, Direct ELISA, Gel Supershift Assay
Purified
NR1H3
Recombinant human LXRGlc alpha (amino acids 2 - 97) and mouse myeloma cells (NS-1)
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10062
Western Blot, Immunoprecipitation, ELISA, Gel Shift
Unconjugated
A liquid formulation in physiologic saline with 0.1% NaN3. with 0.1% Sodium Azide
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, NR1H3, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1
Mouse
IgG2a
100 ug
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Monoclonal
Human
Videos