missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-LTA, Flag/His, Clone: 2B6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004049-M02.100ug
Additional Details : Weight : 0.02000kg
Specifications
LTA | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
LTA | |
LT/TNFB/TNFSF1 | |
Mouse | |
Affinity chromatography | |
RUO | |
4049 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Western Blot | |
2B6 | |
Mouse monoclonal antibody raised against a partial recombinant LTA. | |
PBS with no preservative; pH 7.2 | |
BC034729.1 | |
LTA | |
Recombinant Flag/His fusion protein corresponding to amino acids 58-205 of human LTA. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only