Learn More
LRRC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544096
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LNLALNNIEK IENLEGCEEL AKLDLTVNFI GELSSIKNLQ HNIHLKELFL Sequence homology: Cow: 79%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 79%
LRRC6 Polyclonal antibody specifically detects LRRC6 in Human samples. It is validated for Western Blot.Specifications
LRRC6 | |
Polyclonal | |
Unconjugated | |
LRRC6 | |
CILD19; leucine rich repeat containing 6; leucine rich repeat containing 6 (testis); leucine-rich repeat-containing 6 (testis); leucine-rich repeat-containing protein 6; leucine-rich repeat-containing protein 6; protein tilB homolog; leucine-rich testis-specific protein; LRRC6; Lrtp; Mc2; Protein tilB homolog; testis-specific leucine-rich repeat protein; Tslrp; Unknown (protein for MGC:138048) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23639 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86X45 | |
LRRC6 | |
Synthetic peptide directed towards the N-terminal of human LRRC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |