missing translation for 'onlineSavingsMsg'
Learn More

anti-LRH-1/NR5A2, Clone: H2325, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) PP-H2325-10

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462416

  • £408.22 / 100µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Applications:

Immunoprecipitation, Western Blotting

Specifications

Specifications

LRH-1/NR5A2
H2325
Western Blot, Immunoprecipitation
Purified
NR5A2
Recombinant human LRH-1 (amino acids 161 - 280) and mouse myeloma cells (NS-1)
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2494
Western Blot, Immunoprecipitation
Unconjugated
A liquid formulation in physiologic saline with 0.1% NaN3. with 0.1% Sodium Azide
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1 beta, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH1, LRH-1CPF, NR5A2, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
Mouse
IgG3
100 ug
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Monoclonal
Human
Videos