Learn More
LOX-1/OLR1 Rat anti-Mouse, Clone: 214001, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB15641
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458735
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
LOX-1/OLR1 Monoclonal antibody MAB15641 Specifically detects LOX-1/OLR1 in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
LOX-1/OLR1
214001
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
OLR1
Mouse myeloma cell line NS0-derived recombinant mouse LOX-1/OLR1 Arg60-Ile363 Accession # AAG44998
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4973
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
AAG44998
CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse LOX-1/OLR1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human LOX-1 or human LDL, HDL or VLDL is observed.