missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lman1l Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547805
Code : NEW
Additional Details : Weight : 0.00010kg
Descripción
Reconstitute at 0.2 mg/mL in sterile PBS.
Lman1l Polyclonal antibody specifically detects Lman1l in Rat samples. It is validated for Western Blot.Especificaciones
Lman1l | |
Polyclonal | |
Unconjugated | |
LMAN1L | |
complexin III; ERGIC-53L; ERGIC-53-like protein; ERGIC53-like protein; Ergl; Lectin mannose-binding 1-like; lectin, mannose binding 1 like; lectin, mannose-binding, 1 like; Lman1l; LMAN1-like protein; Protein ERGIC-53-like; slamp; Sublingual acinar membrane protein; UNQ2784/PRO7174 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
300743 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q5FB95 | |
LMAN1L | |
E. coli-derived recombinant rat LMAN1L/SLAMP (aa 33-436) | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |