missing translation for 'onlineSavingsMsg'
Learn More

LILRB5/CD85c/LIR-8 Mouse anti-Human, Clone: 395239, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3065

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469865

  • £286.65 / 100µg

Estimated Shipment date
from Supplier 21-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LILRB5/CD85c/LIR-8 Monoclonal antibody MAB3065 Specifically detects LILRB5/CD85c/LIR-8 in Human samples. It is validated for Western Blotting
Specifications

Specifications

LIR-8/CD85c/LILRB5
395239
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LILRB5
Mouse myeloma cell line NS0-derived recombinant human LILRB5/CD85c/LIR-8 isoform 1 Arg18-His456 Accession # O75023
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10990
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O75023
CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human LILRB5/CD85c/LIR-8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, 2, 3, 4, 5, rhLIR-6, or rhILT11/LILRA5 is observed. In Western blots, less than 5% cross-reactivity with rhILT2, 3, 5, rhLIR-6 or rhILT11/LILRA5 is observed, and no cross-reactivity with rhILT1, 4, or 6.
Human
Videos
Documents

For Research Use Only