Learn More
Lipocalin-1 Mouse anti-Human, Clone: 223908, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1708
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448845
  • £376.32 / 500µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Lipocalin-1 Monoclonal antibody MAB1708 Specifically detects Lipocalin-1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Lipocalin-1
223908
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LCN1
Mouse myeloma cell line NS0-derived recombinant human Lipocalin-1 His19-Asp176 Accession # P31025
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3933
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P31025
LCN1, lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, Lipocalin1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGP, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin)
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Lipocalin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Lipocalin-2.