missing translation for 'onlineSavingsMsg'
Learn More

LINGO-1 Mouse anti-Human, Clone: 332233, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3086

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459885

  • £249.49 / 100µg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

LINGO-1 Monoclonal antibody MAB3086 Specifically detects LINGO-1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

LINGO-1
332233
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LINGO1
Mouse myeloma cell line NS0-derived recombinant human LINGO-1 Thr40-Thr556 Accession # NP_116197
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84894
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_116197
FLJ14594, LERN1, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LINGO1, LRRN6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human LINGO-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) LINGO-2 or rhLINGO-3 is observed.
Human
Videos

For Research Use Only