Learn More
LIM1 Mouse anti-Human, Clone: 320416, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2725
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429655
  • £288.60 / 100µg
Estimated Shipment
14-09-2020
Description
Description
LIM1 Monoclonal antibody MAB2725 Specifically detects LIM1 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
LIM1
320416
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LHX1
E. coli-derived recombinant human LIM1 Leu265-Trp406 Accession # P48742
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3975
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
P48742
hLim-1, Homeobox protein Lim-1, LHX1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human LIM1 in direct ELISAs and Western blots.