missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523966
Code : LD
Additional Details : Weight : 0.01000kg
Description
LILRA2 Polyclonal antibody specifically detects LILRA2 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
LILRA2 | |
Polyclonal | |
Unconjugated | |
LILRA2 | |
CD85 antigen-like family member H; CD85h; ILT1; ILT-1; Immunoglobulin-like transcript 1; leucocyte Ig-like receptor A2; leukocyte immunoglobulin like receptor A2; leukocyte immunoglobulin-like receptor 7; Leukocyte immunoglobulin-like receptor subfamily A member 2; leukocyte immunoglobulin-like receptor subfamily A member 2 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2; LILRA2; LIR7; LIR-7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
11027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8N149 | |
LILRA2 | |
KLH conjugated synthetic peptide between 55-81 amino acids from the central region of human LILRA2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |