Learn More
LIGHT/TNFSF14 Rat anti-Mouse, Clone: 261639, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1794
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488925
  • £275.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
LIGHT/TNFSF14 Monoclonal antibody MAB1794 Specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
LIGHT/TNFSF14
Unconjugated
Purified
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT. Asp72-Val239
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
261639
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Rat
IgG2a
100μg
Primary
Murine
Videos