missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIGHT/TNFSF14 Mouse anti-Human, Clone: 115520, R&D Systems™
Mouse Monoclonal Antibody
Brand: RD Systems (Bio-Techne) MAB664
Code : Z1
Additional Details : Weight : 0.00010kg
Description
LIGHT/TNFSF14 Monoclonal antibody MAB664 Specifically detects LIGHT/TNFSF14 in Human samples. It is validated for ELISA Capture, Flow CytometrySpecifications
LIGHT/TNFSF14 | |
115520 | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
O43557 | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
Mouse | |
IgG1 | |
500 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Monoclonal | |
Detects human LIGHT/TNFSF14 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) Fas Ligand, rhAPRIL, recombinant mouse TRANCE, rhTNF-alpha or rhTRAIL is observed. | |
Human |
Flow Cytometry, CyTOF, ELISA | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Purified | |
TNFSF14 | |
Mouse myeloma cell line NS0-derived recombinant human LIGHT/TNFSF14 Asp74-Val240 Accession # O43557 | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
8740 | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
For Research Use Only